accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHNH2OHO

CAS RN 164332-89-2

IUPAC Name: 3-(4-aminophenyl)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: 3-(4-aminophenyl)-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 280.32
Molecular Formula: C14H20N2O4
InChi Key: NDMVQEZKACRLDP-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC=C(N)C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results