accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2ONaH3CONSOHNOFFNOH3C

CAS RN 164579-32-2

IUPAC Name: sodium 5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methanesulfinyl]-2,3-dihydro-1H-1,3-benzodiazol-1-ide hydrate
Synonyms: sodium 5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methanesulfinyl]-2,3-dihydro-1H-1,3-benzodiazol-1-ide hydrate
Molecular Weight (g/mol): 425.38
Molecular Formula: C16H18F2N3NaO5S
InChi Key: QHNXIKDJFYGZTL-UHFFFAOYNA-N
SMILES: O.[Na+].COC1=CC=NC(CS(=O)C2NC3=CC(OC(F)F)=CC=C3[N-]2)=C1OC
Grade 
  • (1)
  • (5)
Percent Purity 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (5)
  • (5)
  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)