accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1646-26-0

IUPAC Name: 1-(1-benzofuran-2-yl)ethan-1-one
Synonyms: acetylbenzofuran
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: YUTFQTAITWWGFH-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=CC=CC=C2O1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)