accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNOONH3CCH3SH3COO(E)

CAS RN 1646-88-4

IUPAC Name: (E)-(2-methanesulfonyl-2-methylpropylidene)amino N-methylcarbamate
Synonyms: aldicarb sulfone
Molecular Weight (g/mol): 222.26
Molecular Formula: C7H14N2O4S
InChi Key: YRRKLBAKDXSTNC-WEVVVXLNSA-N
SMILES: CNC(=O)O\N=C\C(C)(C)S(C)(=O)=O
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results