accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16466-61-8

IUPAC Name: N'-[(tert-butoxy)carbonyl](tert-butoxy)carbohydrazide
Synonyms: N'-(tert-butoxycarbonyl)tert-butoxycarbohydrazide
Molecular Weight (g/mol): 232.28
Molecular Formula: C10H20N2O4
InChi Key: TYSZETYVESRFNT-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NNC(=O)OC(C)(C)C
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)