accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16466-97-0

IUPAC Name: bromo(prop-1-yn-1-yl)magnesium
Synonyms: bromo(prop-1-yn-1-yl)magnesium
Molecular Weight (g/mol): 143.27
Molecular Formula: C3H3BrMg
InChi Key: YVJBCTFVENNIOV-UHFFFAOYSA-M
SMILES: CC#C[Mg]Br
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)