accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 164666-68-6

IUPAC Name: 6-chloro-2-methylpyridin-3-amine
Synonyms: 6-chloro-2-methylpyridin-3-amine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: QVCIIOZINFCMDJ-UHFFFAOYSA-N
SMILES: CC1=NC(Cl)=CC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results