accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16473-35-1

IUPAC Name: [4-(chloromethyl)phenyl]methanol
Synonyms: [4-(chloromethyl)phenyl]methanol
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: OGALXJIOJZXBBP-UHFFFAOYSA-N
SMILES: OCC1=CC=C(CCl)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)