accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHOCH3O

CAS RN 16475-90-4

IUPAC Name: methyl 5-acetyl-2-hydroxybenzoate
Synonyms: methyl 5-acetyl-2-hydroxybenzoate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: XLSMGNNWSRNTIQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC=C1O)C(C)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)