accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16483-18-4

IUPAC Name: palladium(2+) bis(ethane-1,2-diamine) dichloride
Synonyms: palladium(2+) bis(ethylenediamine) dichloride
Molecular Weight (g/mol): 297.52
Molecular Formula: C4H16Cl2N4Pd
InChi Key: VBDQNUWZQXYUDP-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pd++].NCCN.NCCN
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results