accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CONP

CAS RN 164858-78-0

IUPAC Name: 2-[2-(diphenylphosphanyl)phenyl]-4-(propan-2-yl)-4,5-dihydro-1,3-oxazole
Synonyms: (S)-iPr-PHOX
Molecular Weight (g/mol): 373.44
Molecular Formula: C24H24NOP
InChi Key: OUQSAXROROGQEE-UHFFFAOYNA-N
SMILES: CC(C)C1COC(=N1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results