accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16493-80-4

IUPAC Name: (4R)-4-ethyloctanoate
Synonyms: 4-ethyloctanoic acid
Molecular Weight (g/mol): 171.26
Molecular Formula: C10H19O2
InChi Key: PWKJMPFEQOHBAC-SECBINFHSA-M
SMILES: CCCC[C@@H](CC)CCC([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)