accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CH2NCH3HCl

CAS RN 16496-99-4

IUPAC Name: 1-[1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,9,10,10a-octahydrophenanthren-1-yl]methanamine hydrochloride
Synonyms: 1-(7-isopropyl-1,4a-dimethyl-2,3,4,9,10,10a-hexahydrophenanthren-1-yl)methanamine hydrochloride
Molecular Weight (g/mol): 321.93
Molecular Formula: C20H32ClN
InChi Key: CVPQLGCAWUAYPF-UHFFFAOYNA-N
SMILES: Cl.CC(C)C1=CC2=C(C=C1)C1(C)CCCC(C)(CN)C1CC2
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results