accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCl(E/Z)

CAS RN 16507-61-2

IUPAC Name: 1-chlorooctadec-9-ene
Synonyms: 1-chlorooctadec-9-ene
Molecular Weight (g/mol): 286.93
Molecular Formula: C18H35Cl
InChi Key: IFABLCIRROMTAN-UHFFFAOYSA-N
SMILES: CCCCCCCCC=CCCCCCCCCCl
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)