accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16518-64-2

IUPAC Name: methyl 3-(dimethylamino)benzoate
Synonyms: methyl 3-(dimethylamino)benzoate
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: CABFTHPIDKWPNQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(=C1)N(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results