accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 165190-76-1

IUPAC Name: 4,7-bis(thiophen-2-yl)-2,1,3-benzothiadiazole
Synonyms: 4,7-bis(thiophen-2-yl)-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 300.41
Molecular Formula: C14H8N2S3
InChi Key: XGERJWSXTKVPSV-UHFFFAOYSA-N
SMILES: S1C=CC=C1C1=CC=C(C2=CC=CS2)C2=NSN=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results