accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16532-79-9

IUPAC Name: 2-(4-bromophenyl)acetonitrile
Synonyms: benzeneacetonitrile, 4-bromo-
Molecular Weight (g/mol): 196.05
Molecular Formula: C8H6BrN
InChi Key: MFHFWRBXPQDZSA-UHFFFAOYSA-N
SMILES: BrC1=CC=C(CC#N)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results