accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1655-54-5

OHONHNOONOO

CAS RN 1655-54-5

IUPAC Name: 2-[(2,4-dinitrophenyl)amino]-3-phenylpropanoic acid
Synonyms: 2-[(2,4-dinitrophenyl)amino]-3-phenylpropanoic acid
Molecular Weight (g/mol): 331.28
Molecular Formula: C15H13N3O6
InChi Key: HJQHTLAEPSKXQJ-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=CC=C1)NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)