CAS RN 1655-57-8
CAS RN 1655-57-8
IUPAC Name:
2-[(2,4-dinitrophenyl)amino]-4-methylpentanoic acid
Synonyms:
N-(2,4-dinitrophenyl)leucine
Molecular Weight (g/mol):
297.27
Molecular Formula:
C12H15N3O6
InChi Key:
STMDPCBYJCIZOD-UHFFFAOYNA-N
SMILES:
CC(C)CC(NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O
1
–
1
of
1
results
1
N-(2,4-Dinitrophenyl)-L-leucine 99.0+%, TCI America™
CAS: 1655-57-8 Molecular Formula: C12H15N3O6 Molecular Weight (g/mol): 297.267 MDL Number: MFCD00038102 InChI Key: STMDPCBYJCIZOD-JTQLQIEISA-N Synonym: N-Dnp-L-leucine, Dnp-Leu-OH PubChem CID: 15487857 SMILES: CC(C)CC(C(=O)O)NC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-]