accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1655-68-1

IUPAC Name: 1-methoxy-3-phenoxybenzene
Synonyms: 1-methoxy-3-phenoxybenzene
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: CBVXNDCIOLXDFD-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results