accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1656-44-6

IUPAC Name: 2,4-dinitrobenzene-1-sulfonyl chloride
Synonyms: 2,4-dinitrobenzenesulfonyl chloride
Molecular Weight (g/mol): 266.61
Molecular Formula: C6H3ClN2O6S
InChi Key: SSFSNKZUKDBPIT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C(=C1)[N+]([O-])=O)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)