accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1656-94-6

IUPAC Name: bis[(pyridin-3-yl)methyl]amine
Synonyms: bis(pyridin-3-ylmethyl)amine
Molecular Weight (g/mol): 199.26
Molecular Formula: C12H13N3
InChi Key: FEBQXMFOLRVSGC-UHFFFAOYSA-N
SMILES: C(NCC1=CC=CN=C1)C1=CC=CN=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results