accessibility menu, dialog, popup

UserName

Viewing profile as user:
OCrOOOMg

CAS RN 16569-85-0

IUPAC Name: magnesium(2+) dioxochromiumbis(olate)
Synonyms: magnesium chromate
Molecular Weight (g/mol): 140.3
Molecular Formula: CrMgO4
InChi Key: CRGGPIWCSGOBDN-UHFFFAOYSA-N
SMILES: [Mg++].[O-][Cr]([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results