accessibility menu, dialog, popup

UserName

Viewing profile as user:
FNFFF

CAS RN 16582-93-7

IUPAC Name: 2,3,4,5-tetrafluorobenzonitrile
Synonyms: 2,3,4,5-tetrafluorobenzonitrile
Molecular Weight (g/mol): 175.09
Molecular Formula: C7HF4N
InChi Key: GLTGXIGJLCSEAM-UHFFFAOYSA-N
SMILES: FC1=CC(C#N)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results