accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16588-24-2

BrNOOCl

CAS RN 16588-24-2

IUPAC Name: 4-bromo-1-chloro-2-nitrobenzene
Synonyms: 4-bromo-1-chloro-2-nitrobenzene
Molecular Weight (g/mol): 236.45
Molecular Formula: C6H3BrClNO2
InChi Key: DJRYWPGOQTUJMQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)C=CC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results