accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOBrCl

CAS RN 16588-26-4

IUPAC Name: 2-bromo-1-chloro-4-nitrobenzene
Synonyms: 2-bromo-1-chloro-4-nitrobenzene
Molecular Weight (g/mol): 236.45
Molecular Formula: C6H3BrClNO2
InChi Key: CGTVUAQWGSZCFH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Br)=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results