accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClFFFNCO

CAS RN 16588-69-5

IUPAC Name: 4-chloro-1-isocyanato-2-(trifluoromethyl)benzene
Synonyms: 4-chloro-1-isocyanato-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 221.56
Molecular Formula: C8H3ClF3NO
InChi Key: SBDPJLJFODHSFF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(C=CC(Cl)=C1)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results