accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFNCO

CAS RN 16588-74-2

IUPAC Name: 1-isocyanato-3,5-bis(trifluoromethyl)benzene
Synonyms: 1-isocyanato-3,5-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 255.12
Molecular Formula: C9H3F6NO
InChi Key: NRSSOFNMWSJECS-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(=C1)N=C=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (3)
  • (1)
  • (4)
Boiling Point 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (6)
  • (3)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)