accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 165881-36-7

IUPAC Name: 4-(4-bromobutyl)-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo[6.1.1.0²,⁶]decane
Synonyms: 4-(4-bromobutyl)-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo[6.1.1.0²,⁶]decane
Molecular Weight (g/mol): 315.06
Molecular Formula: C14H24BBrO2
InChi Key: DQGAIMVYOVPBSD-UHFFFAOYNA-N
SMILES: CC1(C)C2CC1C1(C)OB(CCCCBr)OC1C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results