accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 165904-27-8

IUPAC Name: 2-(3,3-diethoxypropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3,3-diethoxypropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 258.17
Molecular Formula: C13H27BO4
InChi Key: VEDSPUPKZZMMLL-UHFFFAOYSA-N
SMILES: CCOC(CCB1OC(C)(C)C(C)(C)O1)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results