accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 1660-04-4

IUPAC Name: 1-(adamantan-1-yl)ethan-1-one
Synonyms: 1-(adamantan-1-yl)ethanone
Molecular Weight (g/mol): 178.28
Molecular Formula: C12H18O
InChi Key: DACIGVIOAFXPHW-UHFFFAOYSA-N
SMILES: CC(=O)C12CC3CC(CC(C3)C1)C2
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (5)