accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COPOPOOCH3H3COCH3CH3OCH3CH3

CAS RN 1660-95-3

IUPAC Name: bis(propan-2-yl) {[bis(propan-2-yloxy)phosphoryl]methyl}phosphonate
Synonyms: diisopropyl (diisopropoxyphosphoryl)methylphosphonate
Molecular Weight (g/mol): 344.33
Molecular Formula: C13H30O6P2
InChi Key: ODTQUKVFOLFLIQ-UHFFFAOYSA-N
SMILES: CC(C)OP(=O)(CP(=O)(OC(C)C)OC(C)C)OC(C)C
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)