accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16605-91-7

IUPAC Name: 2,3-dichloro-1,1'-biphenyl
Synonyms: 2,3-dichlorobiphenyl
Molecular Weight (g/mol): 223.1
Molecular Formula: C12H8Cl2
InChi Key: XOMKZKJEJBZBJJ-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1Cl)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results