accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONNOO

CAS RN 166104-19-4

IUPAC Name: tert-butyl 5-nitro-1H-indole-1-carboxylate
Synonyms: tert-butyl 5-nitroindole-1-carboxylate
Molecular Weight (g/mol): 262.27
Molecular Formula: C13H14N2O4
InChi Key: GSHXPDVMVVLYLH-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C=CC2=CC(=CC=C12)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results