accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16619-55-9

IUPAC Name: 2,2-dibromo-1,3-diphenylpropane-1,3-dione
Synonyms: 2,2-dibromo-1,3-diphenylpropane-1,3-dione
Molecular Weight (g/mol): 382.05
Molecular Formula: C15H10Br2O2
InChi Key: GSWSUDFFJVJMLG-UHFFFAOYSA-N
SMILES: BrC(Br)(C(=O)C1=CC=CC=C1)C(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results