accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSCH3O

CAS RN 16630-52-7

IUPAC Name: 3-(methylsulfanyl)butanal
Synonyms: 3-(methylsulfanyl)butanal
Molecular Weight (g/mol): 118.19
Molecular Formula: C5H10OS
InChi Key: NCBDFIPMWRKPDU-UHFFFAOYNA-N
SMILES: CSC(C)CC=O
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results