accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOSCH3

CAS RN 16630-66-3

IUPAC Name: methyl 2-(methylsulfanyl)acetate
Synonyms: methyl 2-(methylsulfanyl)acetate
Molecular Weight (g/mol): 120.17
Molecular Formula: C4H8O2S
InChi Key: HZYCAKGEXXKCDM-UHFFFAOYSA-N
SMILES: COC(=O)CSC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results