accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOBOHHO

CAS RN 166386-48-7

IUPAC Name: (4-methanesulfinylphenyl)boronic acid
Synonyms: 4-methanesulfinylphenylboronic acid
Molecular Weight (g/mol): 184.02
Molecular Formula: C7H9BO3S
InChi Key: YOTGALZTDVXUKZ-UHFFFAOYNA-N
SMILES: CS(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results