accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNH

CAS RN 1664-40-0

IUPAC Name: N1-phenylethane-1,2-diamine
Synonyms: N1-phenylethane-1,2-diamine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: OCIDXARMXNJACB-UHFFFAOYSA-N
SMILES: NCCNC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)