accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16642-92-5

IUPAC Name: (2E)-3-[4-(trifluoromethyl)phenyl]prop-2-enoate
Synonyms: (2E)-3-[4-(trifluoromethyl)phenyl]prop-2-enoate
Molecular Weight (g/mol): 215.15
Molecular Formula: C10H6F3O2
InChi Key: ANRMAUMHJREENI-ZZXKWVIFSA-M
SMILES: [O-]C(=O)\C=C\C1=CC=C(C=C1)C(F)(F)F
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)