accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16644-30-7

IUPAC Name: 3-(chloromethyl)-2-hydroxy-5-nitrobenzaldehyde
Synonyms: 3-(chloromethyl)-2-hydroxy-5-nitrobenzaldehyde
Molecular Weight (g/mol): 215.59
Molecular Formula: C8H6ClNO4
InChi Key: VPZKJFJWKLYFQD-UHFFFAOYSA-N
SMILES: OC1=C(CCl)C=C(C=C1C=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results