accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNCH3NOO

CAS RN 16648-53-6

IUPAC Name: ethyl N-(4-methyl-3-nitrophenyl)carbamate
Synonyms: ethyl N-(4-methyl-3-nitrophenyl)carbamate
Molecular Weight (g/mol): 224.22
Molecular Formula: C10H12N2O4
InChi Key: ZSEVLTABICCPPU-UHFFFAOYSA-N
SMILES: CCOC(=O)NC1=CC(=C(C)C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results