accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ONHOO

CAS RN 1665-48-1

IUPAC Name: 5-[(3,5-dimethylphenoxy)methyl]-1,3-oxazolidin-2-one
Synonyms: metaxalone
Molecular Weight (g/mol): 221.26
Molecular Formula: C12H15NO3
InChi Key: IMWZZHHPURKASS-UHFFFAOYNA-N
SMILES: CC1=CC(OCC2CNC(=O)O2)=CC(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results