accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOBr

CAS RN 16650-55-8

IUPAC Name: 4-bromonaphthalene-1-carboxylic acid
Synonyms: 4-bromonaphthalene-1-carboxylic acid
Molecular Weight (g/mol): 251.08
Molecular Formula: C11H7BrO2
InChi Key: FIJIPZQZVLCOMB-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=CC2=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)