CAS RN 16652-36-1
IUPAC Name:
2-bromo-3-fluoropropanoic acid
Synonyms:
2-bromo-3-fluoropropanoic acid
Molecular Weight (g/mol):
170.97
Molecular Formula:
C3H4BrFO2
InChi Key:
FBPUCVAAEPPEMG-UHFFFAOYNA-N
SMILES:
OC(=O)C(Br)CF
1
–
1
of
1
results
1
Promotions Available
2-Bromo-3-fluoropropionic acid, 96%, Thermo Scientific Chemicals
2-Bromo-3-fluoropropionic acid, 96%, Thermo Scientific Chemicals
CAS: 16652-36-1 Molecular Formula: C3H4BrFO2 Molecular Weight (g/mol): 170.97 MDL Number: MFCD09800640 InChI Key: FBPUCVAAEPPEMG-UHFFFAOYNA-N Synonym: 2-bromo-3-fluoropropionic acid,acmc-20ao2z,3-fluoro-2-bromo-propionic acid PubChem CID: 316511 IUPAC Name: 2-bromo-3-fluoropropanoic acid SMILES: OC(=O)C(Br)CF