accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3CH3H3C

CAS RN 1667-01-2

IUPAC Name: 1-(2,4,6-trimethylphenyl)ethan-1-one
Synonyms: 1-(2,4,6-trimethylphenyl)ethanone
Molecular Weight (g/mol): 162.23
Molecular Formula: C11H14O
InChi Key: XWCIICLTKWRWCI-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)C=C(C)C=C1C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)