accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1667-04-5

IUPAC Name: 2-chloro-1,3,5-trimethylbenzene
Synonyms: 2-chloro-1,3,5-trimethylbenzene
Molecular Weight (g/mol): 154.64
Molecular Formula: C9H11Cl
InChi Key: WDZACGWEPQLKOM-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(Cl)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results