CAS RN 16681-77-9
IUPAC Name:
1-methyl-1H-1,2,3,4-tetrazole
Synonyms:
1-methyl-1,2,3,4-tetrazole
Molecular Weight (g/mol):
84.08
Molecular Formula:
C2H4N4
InChi Key:
OMAFFHIGWTVZOH-UHFFFAOYSA-N
SMILES:
CN1C=NN=N1
1
–
1
of
1
results
1
1-Methyl-1H-tetrazole 98.0+%, TCI America™
CAS: 16681-77-9 Molecular Formula: C2H4N4 Molecular Weight (g/mol): 84.082 MDL Number: MFCD00129972 InChI Key: OMAFFHIGWTVZOH-UHFFFAOYSA-N PubChem CID: 140123 IUPAC Name: 1-methyltetrazole SMILES: CN1C=NN=N1