accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16694-18-1

IUPAC Name: 4-bromothiophene-2-carboxylic acid
Synonyms: 4-bromothiophene-2-carboxylic acid
Molecular Weight (g/mol): 207.04
Molecular Formula: C5H3BrO2S
InChi Key: HJZFPRVFLBBAMU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Br)=CS1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)