accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOONOSOOCH3SOOH3C

CAS RN 16695-22-0

IUPAC Name: 2-(N-{2-[(4-methylbenzenesulfonyl)oxy]ethyl}4-methylbenzenesulfonamido)ethyl 4-methylbenzene-1-sulfonate
Synonyms: 2-(N-{2-[(4-methylbenzenesulfonyl)oxy]ethyl}4-methylbenzenesulfonamido)ethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 567.69
Molecular Formula: C25H29NO8S3
InChi Key: VJDZYMIEDBLSDT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCCN(CCOS(=O)(=O)C1=CC=C(C)C=C1)S(=O)(=O)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results